2-Methyl-N-(3-oxetanylidene)propane-2-sulfinamide 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Methyl-N-(oxetan-3-ylidene)propane-2-sulphinamide
A105856-100MG
A105856-250MG
A105856-25G
A105856-1G
A105856-5G
A105856-10G
103796-954EA
21
USD
103796-954
103822-700
76809-626
76809-628
76809-630
77309-564
77309-566
77549-018
2-Methyl-N-(3-oxetanylidene)propane-2-sulfinamide 97%
2-Methyl-N-(3-oxetanylidene)propane-2-sulfinamide
Precaución: for Research Use Only
Formula:
C₇H₁₃NO₂S MW: 175.25 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD14702522 CAS Number: 1158098-73-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...