2-Methylmorpholine 97%
Supplier: Ambeed
|
Danger
|
A688166-1G
A688166-25G
A688166-5G
A688166-10G
A688166-250MG
103840-564EA
57
USD
103840-564
76844-782
76844-784
76844-786
77280-756
77280-758
2-Methylmorpholine 97%
2-Methylmorpholine
Precaución: for Research Use Only
Formula:
C₅H₁₁NO MW: 101.1469 g/mol Boiling Pt: 137.1 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD08444368 CAS Number: 27550-90-9 UN: 2734 ADR: 8,3,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...