Methylmalonic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methylmalonic acid

A211160-25G A211160-5G A211160-100G A211160-10G A211160-500G A211160-1000G
103848-780EA 7 USD
103848-780 77087-186 77087-188 77275-152 77275-154 77593-380 77615-794
Methylmalonic acid 97%
Methylmalonic acid

Precaución: for Research Use Only

Formula: C₄H₆O₄
MW: 118.09 g/mol
Boiling Pt: 334.4 °C (760 mm Hg)
Melting Pt: 128…130 °C
Flash Pt: 170.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00002656
CAS Number: 516-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR