2-Methylisonicotinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methylpyridine-4-carboxylic acid

A191406-25G A191406-10G
103816-372EA 8 USD
103816-372 103846-976 77084-812 77111-766
2-Methylisonicotinic acid 98%
2-Methylisonicotinic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₂
MW: 137.14
Boiling Pt: 368.2 °C (760 mm Hg)
Melting Pt: 295…299 °C
Flash Pt: 176.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00513435
CAS Number: 4021-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR