4-Formyl-2-picoline 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methylisonicotinaldehyde

A125510-100G A125510-100MG A125510-250MG A125510-25G A125510-1G A125510-5G A125510-10G
103800-688EA 47 USD
103800-688 76810-866 76810-868 76810-870 76810-872 77298-054 77298-056 77571-522
4-Formyl-2-picoline 98%
4-Formyl-2-picoline

Precaución: for Research Use Only

Formula: C₇H₇NO
MW: 121.1400 g/mol
Boiling Pt: 210.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06410683
CAS Number: 63875-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR