N-Methylphthalimide 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methylisoindoline-1,3-dione

A110961-100G A110961-10G A110961-25G A110961-500G A110961-1000G
103808-056EA 89 USD
103808-056 77257-654 77257-656 77257-658 77257-660 77310-376
N-Methylphthalimide 98%
N-Methylphthalimide

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MW: 161.16 g/mol
Boiling Pt: 286 °C (760 mm Hg)
Melting Pt: 129…132 °C
Flash Pt: 117.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00023063
CAS Number: 550-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR