2-Methylimidazo[1,2-a]pyridine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A222932-1G A222932-5G A222932-10G A222932-250MG
103819-010EA 50 USD
103819-010 103849-888 77088-500 77088-502 77275-518 77275-520
2-Methylimidazo[1,2-a]pyridine-3-carboxylic acid 95%
2-Methylimidazo[1,2-a]pyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.1700 g/mol
Boiling Pt: 170 °C (760 mm Hg)
Melting Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00244615
CAS Number: 21801-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR