2-Methyl-1,3-cyclopentanedione 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methylcyclopentane-1,3-dione

A191905-25G A191905-100G A191905-10G A191905-500G A191905-5G
103847-010EA 10 USD
103847-010 77084-876 77260-514 77260-516 77260-518 77302-270
2-Methyl-1,3-cyclopentanedione 97%
2-Methyl-1,3-cyclopentanedione

Precaución: for Research Use Only

Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 218 °C (760 mm Hg)
Melting Pt: 212…215 °C
Flash Pt: 84.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00001406
CAS Number: 765-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR