2-Methyl-1,3-cyclohexanedione 97%
Supplier: Ambeed
|
Warning
|
A172115-100G
A172115-10G
A172115-500G
A172115-25G
103806-698EA
6
USD
103806-698
103844-982
76769-262
76769-264
76769-266
77082-162
2-Methyl-1,3-cyclohexanedione 97%
2-Methyl-1,3-cyclohexanedione
Precaución: for Research Use Only
Formula:
C₇H₁₀O₂ Boiling Pt: 235.7 °C (760 mm Hg) Melting Pt: 206…208 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001587 CAS Number: 1193-55-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...