Methyl-p-benzoquinone 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methylcyclohexa-2,5-diene-1,4-dione, Methylquinone, Methyl-1,4-benzoquinone

A203456-25G A203456-100G A203456-10G A203456-5G
103848-120EA 5 USD
103848-120 77086-256 77260-854 77260-856 77302-852
Methyl-p-benzoquinone 98%
Methyl-p-benzoquinone

Precaución: for Research Use Only

Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 186.6 °C (760 mm Hg)
Melting Pt: 66…67 °C
Flash Pt: 64.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00001603
CAS Number: 553-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR