2-Methylbenzyl alcohol 98%

Supplier: Ambeed
Danger

Synonyms: 2-(Hydroxymethyl)toluole, (2-Methylphenyl)methanol

A265638-5G A265638-25G
103852-636EA 7 USD
103852-636 77092-336 77304-920
2-Methylbenzyl alcohol 98%
2-Methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 218.4 °C (760 mm Hg)
Melting Pt: 39 °C (760 mm Hg)
Flash Pt: 104.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00004622
CAS Number: 89-95-2
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR