2-Methylbenzophenone 98%

Supplier: Ambeed
Warning

Synonyms: o-Methylbenzophenone

A212067-1000G A212067-25G A212067-5G A212067-100G A212067-10G A212067-500G
103809-470EA 111 USD
103809-470 103848-860 77087-308 77087-310 77087-312 77275-210 77275-212 77275-214
2-Methylbenzophenone 98%
2-Methylbenzophenone

Precaución: for Research Use Only

Formula: C₁₄H₁₂O
MW: 196.25 g/mol
Boiling Pt: 310 °C (760 mm Hg)
Melting Pt: …18 °C
Flash Pt: 119 °C
Storage Temperature: Ambient
MDL Number: MFCD00008518
CAS Number: 131-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR