2-Methylbenzoxazole 95%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-1,3-benzoxazole, 2-Methylbenzo[d]oxazole

A100653-10G A100653-500G A100653-5G A100653-100G A100653-25G
103821-246EA 8 USD
103821-246 77233-038 77233-040 77282-530 77548-892 77548-894
2-Methylbenzoxazole 95%
2-Methylbenzoxazole

Precaución: for Research Use Only

Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 200.5 °C (760 mm Hg)
Melting Pt: 8…10 °C
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00005771
CAS Number: 95-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR