2-Methylbenzenesulfonic acid hydrate 97%
Supplier: Ambeed
|
Warning
|
A236365-250MG
A236365-25G
A236365-1G
103811-084EA
43
USD
103811-084
103850-856
77140-170
77140-172
77304-044
2-Methylbenzenesulfonic acid hydrate 97%
2-Methylbenzenesulfonic acid hydrate
Precaución: for Research Use Only
Formula:
C₇H₁₀O₄S MW: 190.21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29916813 CAS Number: 1914148-59-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...