2-(Methylamino)pyrimidine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methylaminopyrimidine

A488626-100MG A488626-250MG A488626-1G A488626-25G
103828-196EA 55 USD
103828-196 76789-764 76789-766 76843-220 77872-808
2-(Methylamino)pyrimidine 97%
2-(Methylamino)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₇N₃
MW: 109.132 g/mol
Boiling Pt: 216.3 °C (760 mm Hg)
Melting Pt: 60…62 °C
Flash Pt: 84.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00234126
CAS Number: 931-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR