2-Methyl-6-(methylsulfonyl)pyridin-3-amine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-6-(methylsulphonyl)pyridin-3-amine

A474543-100MG A474543-250MG A474543-1G
103828-064EA 339 USD
103828-064 76818-548 76818-550 77307-514
2-Methyl-6-(methylsulfonyl)pyridin-3-amine 95%
2-Methyl-6-(methylsulfonyl)pyridin-3-amine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂S
MW: 186.23 g/mol
Boiling Pt: 441.0±45.0 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD15475086
CAS Number: 897732-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR