2-Methyl-5-phenylthiazole-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A122975-250MG A122975-1G
103800-318EA 193 USD
103800-318 76995-940 77297-908
2-Methyl-5-phenylthiazole-4-carboxylic acid 95%
2-Methyl-5-phenylthiazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₉NO₂S
MW: 219.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04178838
CAS Number: 13743-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR