5-Nitro-2-picoline 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Methyl-5-nitropyridine
A229345-1G
A229345-25G
A229345-5G
A229345-100G
A229345-10G
103819-466EA
9
USD
103819-466
103850-366
77089-198
77089-200
77089-202
77112-926
77112-928
5-Nitro-2-picoline 97%
5-Nitro-2-picoline
Precaución: for Research Use Only
Formula:
C₆H₆N₂O₂ MW: 138.12 g/mol Boiling Pt: 238 °C (760 mm Hg) Melting Pt: 112 °C Flash Pt: 97.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD04114179 CAS Number: 21203-68-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...