2-Methyl-5-nitrophenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-5-nitrophenylboronic acid, pinacol ester, 2-Methyl-5-nitrophenylboronic Acid Pinacol Ester

A838723-1G A838723-25G A838723-5G A838723-100G A838723-10G A838723-250MG
103841-198EA 48 USD
103841-198 76845-432 76845-434 76845-436 77036-624 77036-626 77036-628
2-Methyl-5-nitrophenylboronic acid pinacol ester 98%
2-Methyl-5-nitrophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BNO₄
MW: 263.1 g/mol
Boiling Pt: 378.9 °C (760 mm Hg)
Melting Pt: 90 °C
Flash Pt: 182.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09027081
CAS Number: 957062-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR