5-Nitro-o-toluic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-5-nitrobenzoic acid

A133311-100G A133311-10G A133311-25G A133311-500G A133311-1G A133311-5G
103801-858EA 9 USD
103801-858 77234-602 77234-604 77234-606 77234-608 77298-620 77314-498
5-Nitro-o-toluic acid 98%
5-Nitro-o-toluic acid

Precaución: for Research Use Only

Formula: C₈H₇NO₄
MW: 181.15 g/mol
Melting Pt: 177…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00007371
CAS Number: 1975-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR