2-Methyl-5-nitrobenzaldehyde 98%

Supplier: Ambeed
Warning

A202379-1G A202379-25G A202379-10G A202379-5G
103817-380EA 7 USD
103817-380 103848-020 77086-132 77086-134 77237-204 77302-810
2-Methyl-5-nitrobenzaldehyde 98%
2-Methyl-5-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇NO₃
MW: 165.1500 g/mol
Boiling Pt: 296.4 °C (760 mm Hg)
Melting Pt: 58…61 °C
Flash Pt: 145.287 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03840506
CAS Number: 16634-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR