5-Formyl-2-picoline 97%
Supplier: Ambeed
|
Warning
|
A121165-100G
A121165-25G
A121165-5G
A121165-1G
103800-040EA
7
USD
103800-040
77094-460
77094-462
77297-820
77312-156
5-Formyl-2-picoline 97%
5-Formyl-2-picoline
Precaución: for Research Use Only
Formula:
C₇H₇NO Boiling Pt: 213.7 °C (760 mm Hg) Flash Pt: 87.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08272279 CAS Number: 53014-84-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...