2-Methyl-5-bromopyrimidine-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A361438-1G A361438-250MG
103826-282EA 65 USD
103826-282 103837-296 76840-510 77115-308
2-Methyl-5-bromopyrimidine-4-carboxylic acid 95%
2-Methyl-5-bromopyrimidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O₂
Boiling Pt: 335.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00466678
CAS Number: 100707-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR