2-Methyl-5-(trifluoromethyl)benzoic acid 97%

Supplier: Ambeed
Warning

A522593-25G A522593-5G A522593-10G A522593-250MG A522593-1G
103828-516EA 15 USD
103828-516 103839-630 76843-628 76843-630 77279-788 77279-790 77307-718
2-Methyl-5-(trifluoromethyl)benzoic acid 97%
2-Methyl-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
Boiling Pt: 261 °C (760 mm Hg)
Melting Pt: 123…125 °C
Flash Pt: 111.7 °C
Storage Temperature: Ambient
MDL Number: MFCD01631589
CAS Number: 13055-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR