Carvone 95%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-5-(prop-1-en-2-yl)cyclohex-2-enone

A110097-10G A110097-25G A110097-1G
103797-616EA 9 USD
103797-616 103823-424 76687-484 76687-486 77283-938
Carvone 95%
Carvone

Precaución: for Research Use Only

Formula: C₁₀H₁₄O
MW: 150.2176 g/mol
Boiling Pt: 230.5 °C (760 mm Hg)
Flash Pt: 88.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00062996
CAS Number: 99-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR