2-Methyl-5-pyrimidineboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine, 2-Methylpyrimidine-5-boronic acid pinacol ester

A189276-100MG A189276-250MG A189276-1G A189276-5G
103816-172EA 24 USD
103816-172 103846-766 76813-858 76813-860 77084-530 77084-532
2-Methyl-5-pyrimidineboronic acid pinacol ester 97%
2-Methyl-5-pyrimidineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₇N₂O₂B
MW: 220.0759 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12032615
CAS Number: 1052686-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR