4-Nitro-2-picoline 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-4-nitropyridine

A169671-10G A169671-1G A169671-25G A169671-5G
103806-400EA 5 USD
103806-400 103844-664 76998-174 77081-758 77081-760 77300-992
4-Nitro-2-picoline 98%
4-Nitro-2-picoline

Precaución: for Research Use Only

Formula: C₆H₆N₂O₂
MW: 138.1200 g/mol
Flash Pt: 99.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095077
CAS Number: 13508-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR