2-Methyl-4-nitrophenol 98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-5-nitrotoluene, 4-Nitro-o-cresol

A338821-100G A338821-500G A338821-25G A338821-5G
103836-756EA 16 USD
103836-756 76828-440 76828-442 76839-778 77306-520
2-Methyl-4-nitrophenol 98%
2-Methyl-4-nitrophenol

Precaución: for Research Use Only

Formula: C₇H₇NO₃
Melting Pt: 93…98 °C
Storage Temperature: Ambient
MDL Number: MFCD01734013
CAS Number: 99-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR