4-Methoxy-2-methylbenzaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-4-methoxybenzaldehyde

A440298-1G A440298-25G A440298-5G A440298-100G
103827-658EA 6 USD
103827-658 103838-724 76842-480 76842-482 76842-484 77252-606
4-Methoxy-2-methylbenzaldehyde 98%
4-Methoxy-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 264.1 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD02261771
CAS Number: 52289-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR