4-Fluoro-2-methylphenylboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Fluoro-2-methylbenzeneboronic acid, 2-Methyl-4-fluorophenylboronic acid
A144806-100G
A144806-10G
A144806-500G
A144806-1G
A144806-25G
A144806-5G
103803-454EA
7
USD
103803-454
103841-532
77272-660
77272-662
77272-664
77316-782
77316-784
77316-786
4-Fluoro-2-methylphenylboronic acid 98%
4-Fluoro-2-methylphenylboronic acid
Precaución: for Research Use Only
Formula:
C₇H₈BFO₂ Boiling Pt: 277.4 °C (760 mm Hg) Melting Pt: 191…196 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093072 CAS Number: 139911-29-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...