2-Methyl-4,6-dimethoxypyrimidine 97%

Supplier: Ambeed
Warning

A510463-1G A510463-10G
103828-382EA 13 USD
103828-382 103839-484 76843-472 77116-620
2-Methyl-4,6-dimethoxypyrimidine 97%
2-Methyl-4,6-dimethoxypyrimidine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
MW: 154.169 g/mol
Boiling Pt: 314.4 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06657567
CAS Number: 13566-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR