2-Methyl-4-(trifluoromethoxy)aniline 97%

Supplier: Ambeed
Danger

Synonyms: 2-Amino-5-(trifluoromethoxy)toluene, 4-Trifluoromethoxy-o-toluidine, 4-(Trifluoromethoxy)-2-toluidine

A267881-1G A267881-25G A267881-5G A267881-10G A267881-250MG
103812-886EA 24 USD
103812-886 103852-786 77092-538 77092-540 77092-542 77113-850 77113-852
2-Methyl-4-(trifluoromethoxy)aniline 97%
2-Methyl-4-(trifluoromethoxy)aniline

Precaución: for Research Use Only

Formula: C₈H₈F₃NO
MW: 191.15 g/mol
Boiling Pt: 206.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01631541
CAS Number: 86256-59-9
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR