2-Methyl-4-pyridineboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Methylpyridine-4-boronic acid Pinacol ester, 2-(2-Methyl-4-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A117906-250MG A117906-25G A117906-10G
103798-950EA 15 USD
103798-950 103824-840 77271-314 77271-316 77747-855
2-Methyl-4-pyridineboronic acid pinacol ester 95%
2-Methyl-4-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₈BNO₂
Boiling Pt: 315.9 °C (760 mm Hg)
Melting Pt: 66…71 °C
Flash Pt: 144.8 °C
Storage Temperature: Freezer
MDL Number: MFCD08061590
CAS Number: 660867-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR