2-Methyl-4-(1,1,1,2,3,3,3-heptafluoro-2-propyl)aniline 97%
Supplier: Ambeed
|
Danger
|
A225023-10G
A225023-1G
A225023-25G
103819-150EA
16
USD
103819-150
103850-044
76826-278
77088-716
77088-718
2-Methyl-4-(1,1,1,2,3,3,3-heptafluoro-2-propyl)aniline 97%
2-Methyl-4-(1,1,1,2,3,3,3-heptafluoro-2-propyl)aniline
Precaución: for Research Use Only
Formula:
C₁₀H₈F₇N MW: 275.17 g/mol Boiling Pt: 199.838 °C (760 mm Hg) Flash Pt: 83.354 °C Storage Temperature: Freezer |
MDL Number:
MFCD13185889 CAS Number: 238098-26-5 UN: 3082 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...