3-Nitro-2-picoline 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Methyl-3-nitropyridine
A289443-1G
A289443-25G
A289443-5G
A289443-100G
A289443-10G
103813-830EA
6
USD
103813-830
103834-970
76837-540
76837-542
76837-544
77002-342
77002-344
3-Nitro-2-picoline 98%
3-Nitro-2-picoline
Precaución: for Research Use Only
Formula:
C₆H₆N₂O₂ MW: 138.1240 g/mol Boiling Pt: 213.8 °C (760 mm Hg) Melting Pt: 32…33 °C Flash Pt: 83.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04114137 CAS Number: 18699-87-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...