α-Chloro-2-methyl-3-nitrotoluene 98%
Supplier: Ambeed
|
Danger
|
A269144-25G
A269144-5G
A269144-1G
A269144-250MG
103852-856EA
39
USD
103852-856
77092-614
77092-616
77113-868
77113-870
α-Chloro-2-methyl-3-nitrotoluene 98%
α-Chloro-2-methyl-3-nitrotoluene
Precaución: for Research Use Only
Formula:
C₈H₈ClNO₂ MW: 185.61 g/mol Melting Pt: 43…46 °C Flash Pt: 113 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007165 CAS Number: 60468-54-4 UN: 3261 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...