2-Methyl-1-tetralone 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-3,4-dihydronaphthalen-1(2H)-one

A110769-10G A110769-25G A110769-1G
103797-728EA 19 USD
103797-728 103823-536 76796-814 76796-816 77310-336
2-Methyl-1-tetralone 97%
2-Methyl-1-tetralone

Precaución: for Research Use Only

Formula: C₁₁H₁₂O
MW: 160.22 g/mol
Boiling Pt: 262.3 °C (760 mm Hg)
Melting Pt: 15 °C
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00001690
CAS Number: 1590-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR