3-Amino-2-methylbenzotrifluoride 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-3-(trifluoromethyl)aniline

A201436-100G A201436-10G A201436-1G A201436-500G A201436-25G A201436-5G
103847-932EA 9 USD
103847-932 76691-196 76691-198 76691-200 76691-202 77086-010 77302-752
3-Amino-2-methylbenzotrifluoride 97%
3-Amino-2-methylbenzotrifluoride

Precaución: for Research Use Only

Formula: C₈H₈F₃N
MW: 175.16 g/mol
Boiling Pt: 203 °C (760 mm Hg)
Melting Pt: 38…42 °C
Flash Pt: 96.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00153216
CAS Number: 54396-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR