2-Methyl-3-(methylthio)pyrazine 98%

Supplier: Ambeed
Danger

A173939-100G A173939-10G A173939-25G A173939-5G
103806-888EA 5 USD
103806-888 103845-186 76998-312 76998-314 77082-472 77082-474
2-Methyl-3-(methylthio)pyrazine 98%
2-Methyl-3-(methylthio)pyrazine

Precaución: for Research Use Only

Formula: C₆H₈N₂S
Boiling Pt: 213…214 °C (760 mm Hg)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00014610
CAS Number: 2882-20-4
UN: 3334
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR