2-Methyl-2H-indazol-4-amine 95%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-2-methyl-2H-indazole

A313621-250MG A313621-100MG
103825-012EA 74 USD
103825-012 103835-924 77568-632 77668-386
2-Methyl-2H-indazol-4-amine 95%
2-Methyl-2H-indazol-4-amine

Precaución: For research use only. Keep in a dark place, sealed in dry, at 2…8°C

Formula: C₈H₉N₃
MW: 147.1800 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09870044
CAS Number: 82013-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR