2-Methyl-2-(methylamino)propan-1-ol 95%

Supplier: Ambeed
Warning

Synonyms: N,2-Dimethyl-1-hydroxypropan-2-amine, 1-Hydroxy-2-methyl-2-(methylamino)propane

A180743-100MG A180743-10G A180743-1MG A180743-250MG A180743-25G A180743-1G A180743-5G
103807-554EA 61 USD
103807-554 103845-916 77011-978 77011-980 77011-982 77011-984 77011-986 77083-416 77083-418
2-Methyl-2-(methylamino)propan-1-ol 95%
2-Methyl-2-(methylamino)propan-1-ol

Precaución: for Research Use Only

Formula: C₅H₁₃NO
MW: 103.163 g/mol
Boiling Pt: 162 °C (760 mm Hg)
Flash Pt: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12184529
CAS Number: 27646-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR