2-Methyl-1H-pyrrolo[3,2-b]pyridine 97%

Supplier: Ambeed
Warning

A168291-100MG A168291-250MG
103806-270EA 119 USD
103806-270 76973-890 76973-892
2-Methyl-1H-pyrrolo[3,2-b]pyridine 97%
2-Methyl-1H-pyrrolo[3,2-b]pyridine

Precaución: for Research Use Only

Formula: C₈H₈N₂
MW: 132.1600 g/mol
Boiling Pt: 282.8 °C (760 mm Hg)
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD12923706
CAS Number: 73177-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR