2-Methyl-1H-pyrrole 96%

Supplier: Ambeed
Danger

Synonyms: 2-Methylpyrolle

A490341-1G A490341-5G A490341-10G A490341-250MG A490341-25G
103828-214EA 10 USD
103828-214 103839-286 76843-248 76843-250 77265-574 77265-576 77265-578
2-Methyl-1H-pyrrole 96%
2-Methyl-1H-pyrrole

Precaución: for Research Use Only

Formula: C₅H₇N
Boiling Pt: 147.5 °C (760 mm Hg)
Flash Pt: 47 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02822910
CAS Number: 636-41-9
UN: 1992
ADR: 3,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR