2-Methyl-1H-indol-6-ol 97%
Supplier: Ambeed
|
Warning
|
A177875-100MG
A177875-250MG
A177875-1G
A177875-5G
103807-276EA
209
USD
103807-276
103845-612
76769-460
76769-462
77083-010
77083-012
2-Methyl-1H-indol-6-ol 97%
2-Methyl-1H-indol-6-ol
Precaución: for Research Use Only
Formula:
C₉H₉NO MW: 147.17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09800477 CAS Number: 54584-22-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...