2-Methyl-1H-imidazole-5-carboxylic acid 96%
Supplier: Ambeed
|
Warning
|
A264863-100MG
A264863-250MG
A264863-1G
103812-700EA
52
USD
103812-700
103852-594
77262-316
77262-318
77304-908
2-Methyl-1H-imidazole-5-carboxylic acid 96%
2-Methyl-1H-imidazole-5-carboxylic acid
Precaución: for Research Use Only
Formula:
C₅H₆N₂O₂ MW: 126.11 g/mol Boiling Pt: 475.8 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD00234110 CAS Number: 1457-58-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...