2-Methyl-1H-imidazo[4,5-b]pyridine 98%

Supplier: Ambeed
Warning

A222220-1G A222220-5G A222220-250MG
103818-970EA 82 USD
103818-970 103849-848 77088-450 77088-452 77261-310
2-Methyl-1H-imidazo[4,5-b]pyridine 98%
2-Methyl-1H-imidazo[4,5-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₇N₃
Boiling Pt: 256 °C (760 mm Hg)
Melting Pt: 189…190 °C
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD03673741
CAS Number: 68175-07-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR