2-Methyl-1H-benzoimidazol-5-ylamine 95%

Supplier: Ambeed
Warning

A109407-100G A109407-10G A109407-250MG A109407-1G
103797-504EA 61 USD
103797-504 103823-300 77233-458 77233-460 77233-462 77310-116
2-Methyl-1H-benzoimidazol-5-ylamine 95%
2-Methyl-1H-benzoimidazol-5-ylamine

Precaución: for Research Use Only

Formula: C₈H₉N₃
MW: 147.1800 g/mol
Melting Pt: 95 °C
Flash Pt: 250.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00462859
CAS Number: 29043-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR