2-Methylbenzimidazole 98%

Supplier: Ambeed
Warning

A100835-100G A100835-25G A100835-500G A100835-1000G
103807-772EA 127 USD
103807-772 76809-222 76809-224 76809-226 77296-858
2-Methylbenzimidazole 98%
2-Methylbenzimidazole

Precaución: for Research Use Only

Formula: C₈H₈N₂
Boiling Pt: 339.4 °C (760 mm Hg)
Melting Pt: 175…177 °C
Storage Temperature: Ambient
MDL Number: MFCD00005598
CAS Number: 615-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR