2-Methyl-1,4-phenylene bis(4-((6-(acryloyloxy)hexyl)oxy)benzoate) 95%

Supplier: Ambeed
Warning

A308828-1G A308828-5G A308828-100G A308828-10G A308828-250MG A308828-25G
103814-582EA 11 USD
103814-582 103835-760 76838-526 76838-528 77194-570 77194-572 77194-574 77194-576
2-Methyl-1,4-phenylene bis(4-((6-(acryloyloxy)hexyl)oxy)benzoate) 95%
2-Methyl-1,4-phenylene bis(4-((6-(acryloyloxy)hexyl)oxy)benzoate)

Precaución: for Research Use Only

Formula: C₃₉H₄₄O₁₀
Boiling Pt: 779.2 °C (760 mm Hg)
Flash Pt: 314.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11225140
CAS Number: 125248-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR