2-Methyl-1,4-phenylene bis(4-(((4-(acryloyloxy)butoxy)carbonyl)oxy)benzoate) 97%

Supplier: Ambeed
Warning

A163263-10G A163263-250MG A163263-25G A163263-5G A163263-1G
103805-708EA 19 USD
103805-708 103843-966 77235-928 77235-930 77300-648 77300-650 77320-056
2-Methyl-1,4-phenylene bis(4-(((4-(acryloyloxy)butoxy)carbonyl)oxy)benzoate) 97%
2-Methyl-1,4-phenylene bis(4-(((4-(acryloyloxy)butoxy)carbonyl)oxy)benzoate)

Precaución: for Research Use Only

Formula: C₃₇H₃₆O₁₄
MW: 704.67 g/mol
Boiling Pt: 816.7±65.0 °C (760 mm Hg)
Flash Pt: 330.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD30179603
CAS Number: 187585-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR